accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCl

CAS RN 42217-03-8

IUPAC Name: 1-chlorodocosane
Synonyms: 1-chlorodocosane
Molecular Weight (g/mol): 345.05
Molecular Formula: C22H45Cl
InChi Key: OACXFSZVCDOBKF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)