accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 42235-38-1

IUPAC Name: 1,20-dimethyl icosanedioate
Synonyms: 1,20-dimethyl icosanedioate
Molecular Weight (g/mol): 370.57
Molecular Formula: C22H42O4
InChi Key: PODMJVMUYCFFMK-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCCCCCCCCCCCC(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results