accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 422560-40-5

IUPAC Name: 2-(4-bromophenyl)-N'-hydroxyethanimidamide
Synonyms: 2-(4-bromophenyl)-N'-hydroxyethanimidamide
Molecular Weight (g/mol): 229.08
Molecular Formula: C8H9BrN2O
InChi Key: WHICEIACQGPSMU-UHFFFAOYSA-N
SMILES: NC(CC1=CC=C(Br)C=C1)=NO
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results