CAS RN 42287-94-5
CAS RN 42287-94-5
IUPAC Name:
3-(4-cyanophenyl)propanoic acid
Synonyms:
3-(4-cyanophenyl)propanoic acid
Molecular Weight (g/mol):
175.19
Molecular Formula:
C10H9NO2
InChi Key:
RHIGOXBTMPLABF-UHFFFAOYSA-N
SMILES:
OC(=O)CCC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
Sigma Aldrich 3-(4-Cyanophenyl)propionic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.