CAS RN 4229-38-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sigma Aldrich (Eta5 -Cyclopentadienyl)[N,N-Dimethyl-Nu'-(2-Pyridinylmethylidene)Propane-1,3-Diamine]Ruthenium Hexafluorophosphate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.