accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4229-44-1

IUPAC Name: hydrogen N-methylhydroxylamine chloride
Synonyms: N-methylhydroxylamine
Molecular Weight (g/mol): 83.52
Molecular Formula: CH6ClNO
InChi Key: RGZRSLKIOCHTSI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNO
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)