accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFHO

CAS RN 423-46-1

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexan-1-ol
Synonyms: 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexan-1-ol
Molecular Weight (g/mol): 300.07
Molecular Formula: C6H3F11O
InChi Key: QZFZPVVDBGXQTB-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results