CAS RN 42330-10-9
IUPAC Name:
3-bromoheptan-4-one
Synonyms:
3-bromoheptan-4-one
Molecular Weight (g/mol):
193.08
Molecular Formula:
C7H13BrO
InChi Key:
SFKVBRLKXVRUQW-UHFFFAOYNA-N
SMILES:
CCCC(=O)C(Br)CC
1
–
1
of
1
results
1
Promotions Available
3-Bromo-4-heptanone, 98%
3-Bromo-4-heptanone, 98%
CAS: 42330-10-9 Molecular Formula: C7H13BrO Molecular Weight (g/mol): 193.08 MDL Number: MFCD09757526 InChI Key: SFKVBRLKXVRUQW-UHFFFAOYNA-N Synonym: 3-bromo-4-heptanone,4-heptanone, 3-bromo PubChem CID: 11469593 IUPAC Name: 3-bromoheptan-4-one SMILES: CCCC(=O)C(Br)CC