accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4237-40-5

CH3CH3NOO

CAS RN 4237-40-5

IUPAC Name: 1-(butan-2-yl)-4-nitrobenzene
Synonyms: 1-nitro-4-(sec-butyl)benzene
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: IQRMCUYGEZOTSV-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results