accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHO

CAS RN 42373-30-8

IUPAC Name: 4-amino-3-formylpyridin-1-ium
Synonyms: 4-amino-3-formylpyridin-1-ium
Molecular Weight (g/mol): 123.13
Molecular Formula: C6H7N2O
InChi Key: GTPZHMGXKZIHKW-UHFFFAOYSA-O
SMILES: NC1=CC=[NH+]C=C1C=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)