accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONOSBr

CAS RN 423768-50-7

IUPAC Name: ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
Synonyms: ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
Molecular Weight (g/mol): 302.14
Molecular Formula: C10H8BrNO3S
InChi Key: OIZZMDOBYXRSNQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NOC(=C1)C1=CC=C(Br)S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results