accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 423768-55-2

IUPAC Name: [2-(morpholin-4-yl)pyridin-3-yl]methanol
Synonyms: [2-(morpholin-4-yl)pyridin-3-yl]methanol
Molecular Weight (g/mol): 194.23
Molecular Formula: C10H14N2O2
InChi Key: WESHZTWFDUKWGL-UHFFFAOYSA-N
SMILES: OCC1=C(N=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results