accessibility menu, dialog, popup

UserName

Viewing profile as user:
SH2NNN

CAS RN 423769-74-8

IUPAC Name: 4-(1H-imidazol-1-yl)benzene-1-carbothioamide
Synonyms: 4-(imidazol-1-yl)benzenecarbothioamide
Molecular Weight (g/mol): 203.26
Molecular Formula: C10H9N3S
InChi Key: QUJXJNDFQWCYLF-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(C=C1)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results