CAS RN 42382-46-7
IUPAC Name:
2-(methylsulfanyl)pyrimidin-5-amine
Synonyms:
2-(methylsulfanyl)pyrimidin-5-amine
Molecular Weight (g/mol):
141.19
Molecular Formula:
C5H7N3S
InChi Key:
NRJNAXLXHFQYEX-UHFFFAOYSA-N
SMILES:
CSC1=NC=C(N)C=N1
1
–
1
of
1
results
1
Promotions Available
5-Amino-2-(methylthio)pyrimidine, 96%, Thermo Scientific™
5-Amino-2-(methylthio)pyrimidine, 96%, Thermo Scientific™
CAS: 42382-46-7 Molecular Formula: C5H7N3S Molecular Weight (g/mol): 141.19 MDL Number: MFCD10697095 InChI Key: NRJNAXLXHFQYEX-UHFFFAOYSA-N Synonym: 5-amino-2-methylthio pyrimidine,2-methylthio pyrimidin-5-amine,2-methylsulfanyl pyrimidin-5-amine,2-methylmercapto-5-aminopyrimidine,5-pyrimidinamine,2-methylthio,2-methylsulfanylpyrimidin-5-ylamine,2-methylsulfanyl-5-pyrimidinamine,2-methylthio-5-pyrimidinamine,5-pyrimidinamine, 2-methylthio,5-pyrimidinamine, 2-methylthio-9ci PubChem CID: 12215991 IUPAC Name: 2-methylsulfanylpyrimidin-5-amine SMILES: CSC1=NC=C(N)C=N1