accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 424-18-0

IUPAC Name: methyl 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoate
Synonyms: methyl 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoate
Molecular Weight (g/mol): 328.08
Molecular Formula: C7H3F11O2
InChi Key: NJXMLQHJFDKLKL-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results