accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 424-36-2

IUPAC Name: ethyl 2,2,3,3,4,4,5,5,5-nonafluoropentanoate
Synonyms: ethyl 2,2,3,3,4,4,5,5,5-nonafluoropentanoate
Molecular Weight (g/mol): 292.1
Molecular Formula: C7H5F9O2
InChi Key: JBEYNXOZKKQLOH-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results