accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42409-58-5

IUPAC Name: 5-bromo-3-methoxypyridin-2-amine
Synonyms: 5-bromo-3-methoxypyridin-2-amine
Molecular Weight (g/mol): 203.04
Molecular Formula: C6H7BrN2O
InChi Key: NFBIWMFMHLPVLT-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CN=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results