accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClSClOO

CAS RN 42413-03-6

IUPAC Name: 3-chloro-4-methylbenzene-1-sulfonyl chloride
Synonyms: 3-chloro-4-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 225.08
Molecular Formula: C7H6Cl2O2S
InChi Key: GNYVVCRRZRVBDD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Cl)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results