accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4242-18-6

IUPAC Name: 5,6,7,8-tetrahydronaphthalene-1-carboxylic acid
Synonyms: 5,6,7,8-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: GCFQXKYHWFWGSB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC2=C1CCCC2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)