accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42420-85-9

IUPAC Name: 4,4-diphenylcyclohexan-1-ol
Synonyms: 4,4-diphenylcyclohexan-1-ol
Molecular Weight (g/mol): 252.36
Molecular Formula: C18H20O
InChi Key: UUQQLWRHPNEFIG-UHFFFAOYSA-N
SMILES: OC1CCC(CC1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results