accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOOCH3HCl

CAS RN 4244-84-2

IUPAC Name: ethyl 3-aminopropanoate hydrochloride
Synonyms: Β-alanine ethyl ester hydrochloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: RJCGNNHKSNIUAT-UHFFFAOYSA-N
SMILES: Cl.CCOC(=O)CCN
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (4)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (5)
  • (7)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)