CAS RN 42448-04-4
IUPAC Name:
2,7-diphenyl-9H-carbazole
Synonyms:
2,7-diphenyl-9H-carbazole
Molecular Weight (g/mol):
319.41
Molecular Formula:
C24H17N
InChi Key:
NWLCIOKUOGGKKK-UHFFFAOYSA-N
SMILES:
N1C2=C(C=CC(=C2)C2=CC=CC=C2)C2=C1C=C(C=C2)C1=CC=CC=C1
1
–
1
of
1
results
1
2,7-Diphenyl-9H-carbazole 98.0+%, TCI America™
CAS: 42448-04-4 Molecular Formula: C24H17N Molecular Weight (g/mol): 319.407 InChI Key: NWLCIOKUOGGKKK-UHFFFAOYSA-N PubChem CID: 66717301 IUPAC Name: 2,7-diphenyl-9H-carbazole SMILES: C1=CC=C(C=C1)C2=CC3=C(C=C2)C4=C(N3)C=C(C=C4)C5=CC=CC=C5