accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4245-35-6

IUPAC Name: 3-methylbutyl prop-2-enoate
Synonyms: 3-methylbutyl prop-2-enoate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: ZVYGIPWYVVJFRW-UHFFFAOYSA-N
SMILES: CC(C)CCOC(=O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results