accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4245-76-5

IUPAC Name: N''-methyl-N-nitroguanidine
Synonyms: N''-methyl-N-nitroguanidine
Molecular Weight (g/mol): 118.1
Molecular Formula: C2H6N4O2
InChi Key: XCXKNNGWSDYMMS-UHFFFAOYSA-N
SMILES: CN=C(N)N[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results