accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH

CAS RN 42472-69-5

IUPAC Name: 1-(4-ethynylphenyl)ethan-1-one
Synonyms: 1-(4-ethynylphenyl)ethanone
Molecular Weight (g/mol): 144.17
Molecular Formula: C10H8O
InChi Key: WHFPFLKZXFBCSO-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results