accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4248-19-5

IUPAC Name: tert-butyl carbamate
Synonyms: tert-butyl carbamate
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: LFKDJXLFVYVEFG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(N)=O
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (4)
  • (6)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (4)
  • (4)