accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42492-62-6

IUPAC Name: (2S)-2-[benzyl(methyl)amino]-3-methylbutanoic acid; 4-methylbenzene-1-sulfonic acid
Synonyms: (2S)-2-[benzyl(methyl)amino]-3-methylbutanoic acid; para-toluene sulfonate|N-ME-VAL-OBZL P-TOSYLATE
Molecular Weight (g/mol): 393.5
Molecular Formula: C20H27NO5S
InChi Key: JLGKOCGRFANTSH-YDALLXLXSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)[C@H](N(C)CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)