accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOClCl

CAS RN 4252-78-2

IUPAC Name: 2-chloro-1-(2,4-dichlorophenyl)ethan-1-one
Synonyms: 2-chloro-1-(2,4-dichlorophenyl)ethanone
Molecular Weight (g/mol): 223.48
Molecular Formula: C8H5Cl3O
InChi Key: VYWPPRLJNVHPEU-UHFFFAOYSA-N
SMILES: ClCC(=O)C1=CC=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)