accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNBr

CAS RN 425379-68-6

IUPAC Name: 3-(3-bromophenyl)-1-methyl-1H-pyrazole
Synonyms: 3-(3-bromophenyl)-1-methylpyrazole
Molecular Weight (g/mol): 237.1
Molecular Formula: C10H9BrN2
InChi Key: PNVHBUUFMMMKBR-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results