accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 425379-69-7

IUPAC Name: 5-(3-bromophenyl)-1-methyl-1H-pyrazole
Synonyms: 5-(3-bromophenyl)-1-methylpyrazole
Molecular Weight (g/mol): 237.1
Molecular Formula: C10H9BrN2
InChi Key: RIYUQLBSAMXENJ-UHFFFAOYSA-N
SMILES: CN1N=CC=C1C1=CC(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results