accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42540-40-9

IUPAC Name: sodium (6R,7R)-7-[(2R)-2-(formyloxy)-2-phenylacetamido]-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: cefamandole nafate cephamandole nafate O-formylcefamandole sodium cefamandol nafato
Molecular Weight (g/mol): 512.49
Molecular Formula: C19H17N6NaO6S2
InChi Key: ICZOIXFFVKYXOM-YCLOEFEOSA-M
SMILES: [Na+].CN1N=NN=C1SCC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](OC=O)C1=CC=CC=C1)C2=O)C([O-])=O
Color 
  • (2)
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results