accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFFFFFF

CAS RN 42580-42-7

IUPAC Name: 2,5-bis(trifluoromethyl)benzoic acid
Synonyms: 2,5-bis(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 258.12
Molecular Formula: C9H4F6O2
InChi Key: PINBPLCVZSKLTF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results