accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CO(E)

CAS RN 42588-57-8

IUPAC Name: (2E)-3-ethoxy-2-methylprop-2-enal
Synonyms: (2E)-3-ethoxy-2-methylprop-2-enal
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: KDOAHVPFGIYCEU-AATRIKPKSA-N
SMILES: CCO\C=C(/C)C=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)