accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 426-13-1

IUPAC Name: (1R,3aS,3bS,5S,9aS,9bR,10S,11aS)-1-acetyl-9b-fluoro-1,10-dihydroxy-5,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: fluorometholone fluoro ophtal fluor-op fluoropos isdin brand of fluorometholone allergan brand 1 of fluorometholone allergan brand 3 of fluorometholone pms-fluorometholone fluoromethalone cortilet
Molecular Weight (g/mol): 376.47
Molecular Formula: C22H29FO4
InChi Key: FAOZLTXFLGPHNG-KNAQIMQKSA-N
SMILES: C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(C)=O)[C@@]3(C)C[C@H](O)[C@]2(F)[C@@]2(C)C=CC(=O)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results