accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4265-25-2

IUPAC Name: 2-methyl-1-benzofuran
Synonyms: 2-methylbenzofuran
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: GBGPVUAOTCNZPT-UHFFFAOYSA-N
SMILES: CC1=CC2=CC=CC=C2O1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results