CAS RN 426821-41-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PD 150606
A cell-permeable, non-competitive, selective non-peptide calpain inhibitor [Ki = 210 nM for calpain-1 and 370 nM for calpain-2.