accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNONH2FHCl

CAS RN 426844-23-7

IUPAC Name: 4-fluoropyrrolidine-2-carboxamide hydrochloride
Synonyms: 4-fluoropyrrolidine-2-carboxamide hydrochloride
Molecular Weight (g/mol): 168.6
Molecular Formula: C5H10ClFN2O
InChi Key: YIEACUBSPQGCLW-UHFFFAOYNA-N
SMILES: Cl.NC(=O)C1CC(F)CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results