CAS RN 4271-30-1
IUPAC Name:
2-[(4-aminophenyl)formamido]pentanedioic acid
Synonyms:
2-[(4-aminophenyl)formamido]pentanedioic acid
Molecular Weight (g/mol):
266.25
Molecular Formula:
C12H14N2O5
InChi Key:
GADGMZDHLQLZRI-UHFFFAOYNA-N
SMILES:
NC1=CC=C(C=C1)C(=O)NC(CCC(O)=O)C(O)=O
1
–
1
of
1
results
1
N-(4-Aminobenzoyl)-L-glutamic Acid 97.0+%, TCI America™
CAS: 4271-30-1 Molecular Formula: C12H14N2O5 Molecular Weight (g/mol): 266.253 MDL Number: MFCD00042821 InChI Key: GADGMZDHLQLZRI-VIFPVBQESA-N PubChem CID: 196473 ChEBI: CHEBI:61114 IUPAC Name: (2S)-2-[(4-aminobenzoyl)amino]pentanedioic acid SMILES: C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)N