accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OHNH2CH3

CAS RN 4276-09-9

IUPAC Name: (2R)-2-amino-3-methylbutan-1-ol
Synonyms: (2R)-2-amino-3-methylbutan-1-ol
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: NWYYWIJOWOLJNR-YFKPBYRVSA-N
SMILES: CC(C)[C@@H](N)CO
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)