accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 4276-49-7

IUPAC Name: 1-bromoicosane
Synonyms: 1-bromoeicosane
Molecular Weight (g/mol): 361.45
Molecular Formula: C20H41Br
InChi Key: CZASMUMJSKOHFJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCBr
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)