accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NClCl

CAS RN 42779-56-6

IUPAC Name: 2,4-dichloro-6-methylpyridine
Synonyms: 2,4-dichloro-6-methylpyridine
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: WUGTXQVNSRFDNV-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)