accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4278-43-7

IUPAC Name: zinc(2+) bis(2-methylpropan-2-olate)
Synonyms: zinc(2+) bis(2-methylpropan-2-olate)
Molecular Weight (g/mol): 211.61
Molecular Formula: C8H18O2Zn
InChi Key: DVGVEVPHJQJMPP-UHFFFAOYSA-N
SMILES: [Zn++].CC(C)(C)[O-].CC(C)(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results