accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42783-78-8

IUPAC Name: [(2,2-diethoxyethoxy)methyl]benzene
Synonyms: [(2,2-diethoxyethoxy)methyl]benzene
Molecular Weight (g/mol): 224.3
Molecular Formula: C13H20O3
InChi Key: VTYTZCJKJNWMGA-UHFFFAOYSA-N
SMILES: CCOC(COCC1=CC=CC=C1)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results