accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 427878-70-4

IUPAC Name: ethyl 5-methyl-4-oxo-1H,4H-pyrrolo[2,1-f][1,2,4]triazine-6-carboxylate
Synonyms: ethyl 5-methyl-4-oxo-1H-pyrrolo[2,1-f][1,2,4]triazine-6-carboxylate
Molecular Weight (g/mol): 221.22
Molecular Formula: C10H11N3O3
InChi Key: XCUHVGKOOKXZNK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2NC=NC(=O)C2=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results