accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOH3C

CAS RN 4282-33-1

IUPAC Name: 3,4-dimethyl furan-3,4-dicarboxylate
Synonyms: 3,4-dimethyl furan-3,4-dicarboxylate
Molecular Weight (g/mol): 184.15
Molecular Formula: C8H8O5
InChi Key: SEVRCIAIDGNMFY-UHFFFAOYSA-N
SMILES: COC(=O)C1=COC=C1C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results