accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42826-42-6

IUPAC Name: 2,2-dimethylpropanehydrazide
Synonyms: 2,2-dimethylpropanehydrazide
Molecular Weight (g/mol): 116.16
Molecular Formula: C5H12N2O
InChi Key: OARJXUPBZNUYBG-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NN
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results