CAS RN 4285-48-7
IUPAC Name:
5,6-dibromohex-1-ene
Synonyms:
5,6-dibromohex-1-ene
Molecular Weight (g/mol):
241.95
Molecular Formula:
C6H10Br2
InChi Key:
WFGDPUOTONXJOS-UHFFFAOYNA-N
SMILES:
BrCC(Br)CCC=C
1
–
1
of
1
results
1
5,6-Dibromo-1-hexene 96.0+%, TCI America™
CAS: 4285-48-7 Molecular Formula: C6H10Br2 Molecular Weight (g/mol): 241.954 MDL Number: MFCD00041698 InChI Key: WFGDPUOTONXJOS-UHFFFAOYSA-N PubChem CID: 2755398 IUPAC Name: 5,6-dibromohex-1-ene SMILES: C=CCCC(CBr)Br