accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42854-62-6

H3CNH2OOCH3SOHOO

CAS RN 42854-62-6

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl 2-aminopropanoate
Synonyms: benzyl 2-aminopropanoate; para-toluene sulfonate
Molecular Weight (g/mol): 351.42
Molecular Formula: C17H21NO5S
InChi Key: NWOPHJSSBMABBD-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CC(N)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)