accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClOOH3CNOO

CAS RN 42855-00-5

IUPAC Name: (4,5-dimethoxy-2-nitrophenyl)methyl carbonochloridate
Synonyms: (4,5-dimethoxy-2-nitrophenyl)methyl carbonochloridate
Molecular Weight (g/mol): 275.64
Molecular Formula: C10H10ClNO6
InChi Key: RWWPKIOWBQFXEE-UHFFFAOYSA-N
SMILES: COC1=CC(COC(Cl)=O)=C(C=C1OC)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results