accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3OHNOHOCH3OH3C

CAS RN 42864-78-8

IUPAC Name: hydrogen 1-{[2-(3,4-dimethoxyphenyl)ethyl]amino}-3-(3-methylphenoxy)propan-2-ol chloride
Synonyms: bevantolol
Molecular Weight (g/mol): 381.9
Molecular Formula: C20H28ClNO4
InChi Key: FJTKCFSPYUMXJB-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC1=CC=C(CCNCC(O)COC2=CC=CC(C)=C2)C=C1OC
Concentration 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results